Clidinium Bromide, Reference Standards
About this item
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Certifications: US Pharmacopea Reference Standard
2 Options Available Below
Product Details & Documents
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Certifications: US Pharmacopea Reference Standard
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- 1135000.
- 1135021.
- Description:
- Clidinium Bromide
- Clidinium Bromide Related Compound A
- Synonym(s):
- 3-Hydroxy-1-methylquinuclindinium Bromide
- CAS No.:
- 3485-62-9
- 2096336-00-2
Specifications
Product Family Options
Product Information
- StandardsPack typePkAvailabilityPrice
- 1135000.Clidinium BromideSpecifications:
More
Specifications
Cat. No.1135000.DescriptionClidinium BromideSynonym(s)CAS No.3485-62-9 - 1135021.Clidinium Bromide Related Compound AClidinium Bromide Related Compound AUSP reference standardVial250 mgLog in for availability1 * 250 mg1 360,00 €Specifications:
More
Specifications
Cat. No.1135021.DescriptionClidinium Bromide Related Compound ASynonym(s)3-Hydroxy-1-methylquinuclindinium BromideCAS No.2096336-00-2



