Order Entry
Export
ContactUsLinkComponent
Cefprozil, Reference Standards
Cefprozil, Reference Standards
Supplier:  USP
Certificates
Cefprozil, Reference Standards
Supplier:  USP

Specifications

  • CAS No.:
    110764-35-7
  • Synonym(s):
    (6R,7R)-7-[(S)-2-amino-2-(4-hydroxyphenyl)acetamido]-8-oxo-3-[(Z)-prop-1-enyl]-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid
  • Description:
    Cefprozil L-Epimer

Specifications