Order Entry
Export
Orders LinkContactUsLinkComponent
 

Carglumic Acid, Reference Standards

Supplier:   USP
Carglumic Acid, Reference Standards
Supplier:  USP
Certificates

2 Options Available Below

Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certifications: US Pharmacopea Reference Standard

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • 1096510.
  • 1096586.
  • Description:
  • Carglumic Acid Related Compound A
  • Carglumic Acid
  • Synonym(s):
  • (S)-3-(2,5-dioxoimidazolidin-4-yl)propanoic acid
  • CAS No.:
  • 17027-50-8
  • 1188-38-1

Specifications

Product Family Options

Product Information

  • Standards
    Pack type
    Pk
    Availability
    Price
  • 1096510.
    Carglumic Acid Related Compound A
    Carglumic Acid Related Compound A
    USP reference standard
    Vial
    30 mg
    Log in for availability
    Log in for availability
    1 * 30 mg
    1 * 30 mg - 860,00 €
    860,00 €
    1 * 30 mg
       860,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1096510.
    Description
    Carglumic Acid Related Compound A
    Synonym(s)
    (S)-3-(2,5-dioxoimidazolidin-4-yl)propanoic acid
    CAS No.
    17027-50-8
  • 1096586.
    Carglumic Acid
    Carglumic Acid
    USP reference standard
    Vial
    200 mg
    Log in for availability
    Log in for availability
    1 * 200 mg
    1 * 200 mg - 382,00 €
    382,00 €
    1 * 200 mg
       382,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1096586.
    Description
    Carglumic Acid
    Synonym(s)
    CAS No.
    1188-38-1