Order Entry
Export
Orders LinkContactUsLinkComponent
Caprylic Acid, Reference Standard

Caprylic Acid, Reference Standard

Caprylic Acid, USP reference standard, Vial, 300 mg
Catalog # 1091040.
Supplier:   USP
Caprylic Acid, Reference Standard
Catalog # 1091040.
Supplier:  USP
CAS Number:  
Caprylic Acid, Reference Standard

Caprylic Acid, USP reference standard, Vial, 300 mg

CAS Number:  
Price
391,00 €
Log in for availability
1 * 300 mg
Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certifications: US Pharmacopea Reference Standard

Documents

Specifications

Specifications

  • Pack type:
    Vial
  • Pk:
    300 mg
  • Standards:
    USP reference standard
  • Description:
    Caprylic Acid
  • UN No.:
    3265
  • Cat. No.:
    1091040.
  • Formula:
    C₈H₁₆O₂
  • Description:
    Caprylic Acid
  • CAS No.:
    124-07-2

Specifications