Order Entry
Export
Orders LinkContactUsLinkComponent
Atenolol, Reference Standards

Atenolol, Reference Standards

Supplier:   USP
Atenolol, Reference Standards
Supplier:  USP
Certificates

2 Options Available Below

Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certifications: US Pharmacopea Reference Standard

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • 1044403.
  • 1044367.
  • Description:
  • Atenolol
  • Atenolol
  • Synonym(s):
  • 2,2'-[{[(Isopropylazanediyl)bis(2-hydroxypropane-3,1-diyl)]bis(oxy)}bis(4,1-phenylene)]diacetamide
  • CAS No.:
  • 29122-68-7
  • 87619-83-8
  • Formula:
  • C₁₄H₂₂N₂O₃

Specifications

Product Family Options

Product Information

  • Standards
    Pack type
    Pk
    Availability
    Price
  • 1044403.
    Atenolol
    Atenolol
    USP reference standard
    Vial
    200 mg
    Log in for availability
    Log in for availability
    1 * 200 mg
    1 * 200 mg - 382,00 €
    382,00 €
    1 * 200 mg
       382,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1044403.
    Description
    Atenolol
    Synonym(s)
    CAS No.
    29122-68-7
    Formula
    C₁₄H₂₂N₂O₃
  • 1044367.
    Atenolol
    Atenolol
    USP reference standard
    Vial
    25 mg
    Log in for availability
    Log in for availability
    1 * 25 mg
    1 * 25 mg - 1 130,00 €
    1 130,00 €
    1 * 25 mg
       1 130,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1044367.
    Description
    Atenolol
    Synonym(s)
    2,2'-[{[(Isopropylazanediyl)bis(2-hydroxypropane-3,1-diyl)]bis(oxy)}bis(4,1-phenylene)]diacetamide
    CAS No.
    87619-83-8
    Formula