Atazanavir, Reference Standards
About this item
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Atazanavir System Suitability Mixture: A mixture of atazanavir sulfate, atazanavir R, S, S, S,-diastereomer and atazanavir S,R,S,S diastereomer
Certifications: US Pharmacopea Reference Standard
3 Options Available Below
Product Details & Documents
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Atazanavir System Suitability Mixture: A mixture of atazanavir sulfate, atazanavir R, S, S, S,-diastereomer and atazanavir S,R,S,S diastereomer
Certifications: US Pharmacopea Reference Standard
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- 1044345.
- 1044334.
- 1044356.
- Description:
- Atazanavir System Suitability Mixture
- Atazanavir Sulfate
- Atazanavir Related Compound A
- Synonym(s):
- (S)-2-[(Methoxycarbonyl)amino]-3,3-dimethylbutanoic acid
- CAS No.:
- 229975-97-7
- 162537-11-3
- Formula:
- C₃₈H₅₂N₆O₇, H₂O₄S
Specifications
Product Family Options
Product Information
- StandardsPack typePkAvailabilityPrice
- 1044345.Atazanavir System Suitability MixtureAtazanavir System Suitability MixtureUSP reference standardVial25 mgLog in for availability1 * 25 mg1 370,00 €Specifications:
More
Specifications
Cat. No.1044345.DescriptionAtazanavir System Suitability MixtureSynonym(s)CAS No.Formula - 1044334.Atazanavir SulfateSpecifications:
More
Specifications
Cat. No.1044334.DescriptionAtazanavir SulfateSynonym(s)CAS No.229975-97-7FormulaC₃₈H₅₂N₆O₇, H₂O₄S - 1044356.Atazanavir Related Compound AAtazanavir Related Compound AUSP reference standardVial25 mgLog in for availability1 * 25 mg1 150,00 €Specifications:
More
Specifications
Cat. No.1044356.DescriptionAtazanavir Related Compound ASynonym(s)(S)-2-[(Methoxycarbonyl)amino]-3,3-dimethylbutanoic acidCAS No.162537-11-3Formula



