Argatroban, Reference Standards
About this item
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Certifications: US Pharmacopea Reference Standard
4 Options Available Below
Product Details & Documents
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Certifications: US Pharmacopea Reference Standard
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- 1042408.
- 1042419.
- 1042420.
- 1042437.
- Description:
- Argatroban
- Argatroban Related Compound A
- Argatroban Related Compound B
- Argatroban Related Compound C
- Synonym(s):
- (2R,4R)-1-[N8-nitro- N2-(3-methylquinoline-8-sulfonyl)-L-arginyl]-4-methylpiperidine-2-carboxylic acid
- Ethyl (4R)-1-[N^8-nitro-L-arginyl]-4-methylpiperidine-2-carboxylate dihydrochloride
- (2R,4R)-1-[N^8-amino- N^2-(3-methyl-1,2,3,4-tetrahydroquinoline-8-sulfonyl)-L-arginyl]-4-methylpiperidine-2-carboxylic acid
- CAS No.:
- 141396-28-3
- 74874-10-5
Specifications
Product Family Options
Product Information
- StandardsPack typePkAvailabilityPrice
- 1042408.ArgatrobanSpecifications:
More
Specifications
Cat. No.1042408.DescriptionArgatrobanSynonym(s)CAS No.141396-28-3 - 1042419.Argatroban Related Compound AArgatroban Related Compound AUSP reference standardVial15 mgLog in for availability1 * 15 mg1 390,00 €Specifications:
More
Specifications
Cat. No.1042419.DescriptionArgatroban Related Compound ASynonym(s)(2R,4R)-1-[N8-nitro- N2-(3-methylquinoline-8-sulfonyl)-L-arginyl]-4-methylpiperidine-2-carboxylic acidCAS No.74874-10-5 - 1042420.Argatroban Related Compound BArgatroban Related Compound BUSP reference standardVial20 mgLog in for availability1 * 20 mg1 330,00 €Specifications:
More
Specifications
Cat. No.1042420.DescriptionArgatroban Related Compound BSynonym(s)Ethyl (4R)-1-[N^8-nitro-L-arginyl]-4-methylpiperidine-2-carboxylate dihydrochlorideCAS No. - 1042437.Argatroban Related Compound CArgatroban Related Compound CUSP reference standardVial20 mgLog in for availability1 * 20 mg1 400,00 €Specifications:
More
Specifications
Cat. No.1042437.DescriptionArgatroban Related Compound CSynonym(s)(2R,4R)-1-[N^8-amino- N^2-(3-methyl-1,2,3,4-tetrahydroquinoline-8-sulfonyl)-L-arginyl]-4-methylpiperidine-2-carboxylic acidCAS No.



