
DTBP (Dimethyl 3,3′-dithiopropionimidate dihydrochloride), Sigma-Aldrich®
Catalog # SIALD2388-100MG
Supplier: Merck
CAS Number:
Specifications
- Pack type:Poly bottle
- Pk:100 mg
- Seq:2440
Specifications
About this item
:
:309.28 g/mol
:M-Clarity™ quality level = MQ100
:GHS07
Storage Temperature:Refrigerator
CAS Number:38285-78-8
Formula:C₈H₁₆N₂O₂S₂·2HCl
MDL Number:MFCD00043245