Order Entry
Export
ContactUsLinkComponent
DTBP (Dimethyl 3,3′-dithiopropionimidate dihydrochloride), Pierce™
Irritant
DTBP (Dimethyl 3,3′-dithiopropionimidate dihydrochloride), Pierce™
Catalog # PIER20665
CAS Number:  
undefined
Irritant
DTBP (Dimethyl 3,3′-dithiopropionimidate dihydrochloride), Pierce™
Catalog # PIER20665
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Product Line:
    Pierce™
  • Labeling Method:
    Chemical Labeling
  • Crosslinker Type:
    Homobifunctional
  • Reactive Moiety:
    Imidoester
  • Spacer:
    Cleavable, Mid-Length
  • Form:
    Powder
  • Quantity:
    1 g
  • Type:
    Crosslinker
  • Solubility:
    Water

Specification Test Results

About this item

 :
 :309.28 g/mol
 :GHS07
Storage Temperature:Refrigerator
CAS Number:38285-78-8
Formula:C₈H₁₆N₂O₂S₂·2HCl
MDL Number:MFCD00043245