DTSSP (3,3'-Dithiobis(sulphosuccinimidyl propionate)), Pierce™
PIER21578
:
- Pk:50 mg
- Seq:2370
- Product Line:Pierce™
- Labeling Method:Chemical Labeling
- Crosslinker Type:Homobifunctional
- Reactive Moiety:Sulfo-NHS Ester
- Spacer:Cleavable, Long
- Form:Powder
- Quantity:50 mg
- Type:Crosslinker
- Solubility:Water
:
:564.542 g/mol
:Refrigerator
:81069-02-5
:C₁₄H₁₆N₂O₁₄S₄
:MFCD01861949