Step shelves, white PVC
Catalog # TRNT50109
Supplier: TRIPP
Specifications
- Description:Step shelf
- Seq:03
- W×D×H:457×117×254 mm
- Pk:1
- Weight:0,997903 kg
Specifications
About this item
Step shelves 'step over' bench top supplies, gas valves and outlets. Shelves create extra storage and make work areas easier to clean by lifting up clutter. With no bottom and a cut away back, bottles, supplies and power cords are available under the shelf. Durable PVC construction makes these shelves resistant, easy to clean and move. Built to last, these shelves can be repurposed as needed to give a long performance life.
- Bottomless shelves fit over bench top protrusions
- Open back to allow air flow and cords to pass through
- Safely lip keeps items secure
- 6 sizes and shapes to fit in any space
- White PVC construction
- Durable and easy to clean
- In stock and ready to ship
- American made by TrippNT
- Total replacement lifetime guarantee