DSP (Dithiobis(succinimidyl propionate)), powder, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:57757-57-0
:
Formula:C₁₄H₁₆N₂O₈S₂
MDL Number:MFCD00042045
:404.41 g/mol
:M-Clarity™ quality level = MQ200
Storage Temperature:Freezer
Specification Test Results
- assay:≥97.0% (N)