DSP (Dithiobis(succinimidyl propionate)), Premium Grade, Pierce™
Catalog # PIERPG82081
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:1 mg
- Seq:2070
Specifications
Specification Test Results
- 1 g Sufficient For:2.47L of typical 1X (1mM) reaction solution
- Product Line:Pierce™
- Labeling Method:Chemical Labeling
- Crosslinker Type:Homobifunctional
- Reactive Moiety:NHS Ester
- Spacer:Cleavable, Long
- Form:Solid
- Quantity:1 g / 10 g
- Type:Crosslinker
- Solubility:DMF, DMSO
Specification Test Results
About this item
:
:404.41 g/mol
Storage Temperature:Refrigerator
CAS Number:57757-57-0
Formula:C₁₄H₁₆N₂O₈S₂
MDL Number:MFCD00042045