

Temozolomide
ACRO466760010
:
- Pack type:Glass bottle
- Pk:1 g
- Seq:3047
- Appearance:White to light pink to brown powder
- Infrared spectrum:Conforms
- Loss on drying:≤ 1%
- HPLC:≥ 97.5%
:
:194.15 g/mol
:13.9218
:GHS07
:GHS08
:Refrigerator
:85622-93-1
:C₆H₆N₆O₂
:MFCD00866492