Order Entry
Export
ContactUsLinkComponent
Proflavine hydrochloride (3,6-diaminoacridine hydrochloride)
Irritant
Proflavine hydrochloride (3,6-diaminoacridine hydrochloride)
Catalog # ACRO465860050
CAS Number:  
Irritant
Proflavine hydrochloride (3,6-diaminoacridine hydrochloride)
Catalog # ACRO465860050
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Appearance:
    Dark orange to red-brown powder
  • Infrared spectrum:
    Conforms
  • Titration Argentometry:
    13.6 to 15.0% (Cl)
  • HPLC:
    ≥ 94%
  • Loss on drying:
    ≤ 4% (105°C, 1hr)
  • Solubility:
    (1mg/ml in water) Clear solution

Specification Test Results

About this item

 :
 :245.71 g/mol
Melting Pt:270°C
EINECS:213-469-9
 :GHS07
Storage Temperature:Ambient
CAS Number:952-23-8
Formula:C₁₃H₁₁N₃·HCl
MDL Number:MFCD00066867