
Hydrocortisone 17-Butyrate
Catalog # ACRO465050010
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to almost white crystalline powder
- Infrared spectrum:Conforms
- Loss on drying:≤ 1%
- HPLC:≥ 96.0%
- Specific optical rotation:+47° to +54° (20°C, 589nm) (0.1% in CHCl3)
Specification Test Results
About this item
:
:432.56 g/mol
Merck Index:13.4808
EINECS:237-093-4
:GHS07
Storage Temperature:Ambient
CAS Number:13609-67-1
Formula:C₂₅H₃₆O₆
MDL Number:MFCD00083364