Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Aluminum etch
Aluminum etch (16:1:1:2), CMOS, J.T.Baker®
Aluminum etch (16:1:1:2), CMOS, J.T.Baker®
Aluminum etch (16:1:1:2), CMOS, J.T.Baker®
Catalog # 5465-05
Supplier:
Avantor
CAS Number:
Expand image
Aluminum etch (16:1:1:2), CMOS, J.T.Baker®
Catalog # 5465-05
Supplier:
Avantor
CAS Number:
Specifications
Pk:
3,8 L
Seq:
5138
Specifications
About this item
Product Details
:
Product Details