


Ammonium peroxodisulphate (APS) ACS
Catalog # SIAL248614-100G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Poly bottle
- Pk:100 g
- Seq:3401
Specifications
Specification Test Results
- Assay:98.0%
Specification Test Results
About this item
:
:228.2 g/mol
Merck Index:13,00545
Melting Pt:120 °C
Density:1.98 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-786-5
UN:1444
ADR:5.1,III
:GHS03
:GHS08
:GHS07
CAS Number:7727-54-0
Formula:(NH₄)₂S₂O₈
MDL Number:MFCD00003390