

Dodecamolybdophosphoric acid hydrate ACS
Catalog # SIAL221856-100G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Plastic bottle, wide mouth
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:1843.27 g/mol
Merck Index:12,07498
Melting Pt:78…90 °C
Density:1.62 g/cm³ (25 °C)
Storage Temperature:Ambient
EINECS:234-713-5
UN:3260
ADR:8,III
:GHS03
:GHS05
CAS Number:51429-74-4
Formula:H₃[P(Mo₃O₁₀)₄]·nH₂O
MDL Number:MFCD00149913