Order Entry
Export
ContactUsLinkComponent
Diammonium hydrogen orthophosphate ≥98% ACS
Diammonium hydrogen orthophosphate ≥98% ACS
Catalog # SIAL215996-500G
Supplier:  Merck
CAS Number:  
Diammonium hydrogen orthophosphate ≥98% ACS
Catalog # SIAL215996-500G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle, wide mouth
  • Pk:
    500 g
  • Seq:
    3645

Specifications

Specification Test Results

  • Assay:
    98%

Specification Test Results

About this item

 :
 :132.06 g/mol
Merck Index:13,00546
Melting Pt:185 °C
Density:1.619 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-987-8
REACH:01-2119490974-22
CAS Number:7783-28-0
Formula:(NH₄)₂HPO₄
MDL Number:MFCD00010891