iFSP1 ≥98%
Supplier: Cayman Chemical
Jump to:
About this item
iFSP1 is an inhibitor of apoptosis inducing factor mitochondria-associated 2/ferroptosis suppressor protein 1 (AIFM2/FSP1)
Caution: 98
Synonyms: 1-amino-3-(4-methylphenyl)-pyrido[1,2-a]benzimidazole-2,4-dicarbonitrile, 1-Amino-3-p-tolyl-benzo[4,5]imidazo[1,2-a]pyridine-2,4-dicarbonitrile
- CAS Number:150651-39-1
- Formula:C₂₀H₁₃N₅
- MDL Number:MFCD01572665
- Molecular Weight:323.4 g/mol
- Storage Temperature:Freezer
4 Options Available Below
Product Details & Documents
iFSP1 is an inhibitor of apoptosis inducing factor mitochondria-associated 2/ferroptosis suppressor protein 1 (AIFM2/FSP1)
Caution: 98
Synonyms: 1-amino-3-(4-methylphenyl)-pyrido[1,2-a]benzimidazole-2,4-dicarbonitrile, 1-Amino-3-p-tolyl-benzo[4,5]imidazo[1,2-a]pyridine-2,4-dicarbonitrile
- CAS Number:150651-39-1
- Formula:C₂₀H₁₃N₅
- MDL Number:MFCD01572665
- Molecular Weight:323.4 g/mol
- Storage Temperature:Freezer



