Order Entry
Export
ContactUsLinkComponent
Anti-CD5 Mouse Antibody (APC (Allophycocyanin)) [clone: UCHT2]
Anti-CD5 Mouse Antibody (APC (Allophycocyanin)) [clone: UCHT2]
Supplier:  Tonbo Biosciences
Certificates
undefined
Anti-CD5 Mouse Antibody (APC (Allophycocyanin)) [clone: UCHT2]
Supplier:  Tonbo Biosciences

Specifications

  • Catalog No:
  • 20-0059-T100
  • 20-0059-T025
  • Antigen symbol:
  • CD5
  • CD5
  • Antigen name:
  • CD5 molecule
  • CD5 molecule
  • Antigen synonyms:
  • CD5|LEU1|Lymphocyte antigen T1/Leu-1|T-cell surface glycoprotein CD5|T1
  • CD5|LEU1|Lymphocyte antigen T1/Leu-1|T-cell surface glycoprotein CD5|T1
  • Antibody type:
  • Primary
  • Primary
  • Conjugation:
  • APC (Allophycocyanin)
  • APC (Allophycocyanin)
  • Clone:
  • UCHT2
  • UCHT2
  • Reactivity:
  • Marmoset, Capuchin monkey, Chimpanzee, Human
  • Marmoset, Capuchin monkey, Chimpanzee, Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 921
  • 921
  • Isotype:
  • IgG1
  • IgG1
  • Flow cytometry:
  • Yes
  • Yes
  • Neutralisation:
  • Yes
  • Yes
  • Storage temperature:
  • Store undiluted at 2 - 8 °C, protect from light, do not freeze.
  • Store undiluted at 2 - 8 °C, protect from light, do not freeze.
  • Tested applications:
  • Flow Cytometry
  • Flow Cytometry

Specifications