



Sodium borohydride ≥97%, AnalaR NORMAPUR® for analysis
Catalog # 27883.180
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pk:100 g
- Seq:3401
Specifications
Specification Test Results
- Assay:min. 97%
- Identity:passes test
- Appearance:white hygroscopic crystals
- Chloride (Cl):max. 0,5%
- Sulphate (SO4):max. 0,01%
- Cadmium (Cd):max. 0,005%
- Copper (Cu):max. 0,005%
- Iron (Fe):max. 0,005%
- Lead (Pb):max. 0,005%
- Zinc (Zn):max. 0,005%
Specification Test Results
About this item
:
:37.83 g/mol
Storage Temperature:Ambient
EINECS:241-004-4
UN:1426
ADR:4.3,I
:GHS02
:GHS06
:GHS05
:GHS08
CAS Number:16940-66-2
Formula:NaBH₄
MDL Number:MFCD00003518