Anti-GBA rabbit polyclonal antibody
NOVUNBP1-32271
- Antibody type:Primary
- Antigen name:Glucosylceramidase
- Antigen symbol:GBA
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Zebrafish,Human
- Western blot:Yes
- Gene ID:2629
- Antigen synonyms:glucosylceramidase|alglucerase|Acid beta-glucosidase|GC|GBA1|beta|lysosomal glucocerebrosidase|GLUC|glucosidase|GCB|acid (includes glucosylceramidase)|D-glucosyl-N-acylsphingosine glucohydrolase|imiglucerase|Beta-glucocerebrosidase|acid|EC 3.2.1.45
- Storage buffer:0,1 M Tris (pH 7,0), 0,1 M Glycine and 10% Glycerol
- Storage temperature:Aliquot and store at −20 or −80 °C. Avoid freeze-thaw cycles.
- Concentration:0,82 mg/ml
- Immunogen:Recombinant protein encompassing a sequence within the C-terminus region of human GBA. The exact sequence is proprietary.
- Purification:Immunogen affinity purified
- Pk:0,1 mL
The Glucosylceramidase / GBA Antibody from Novus Biologicals is a rabbit polyclonal antibody to Glucosylceramidase / GBA. This antibody reacts with human, zebrafish. The Glucosylceramidase / GBA Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.
Recommended Dilutions: Immunohistochemistry (Parafin): 1:100-1:500
Type: Primary
Antigen: GBA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Zebrafish