Anti-PTPRC Mouse Antibody (PerCP (Peridinin-Chlorophyll Protein Complex)-Cy5.5®) [clone: HI30]
Catalog # 65-0459-T025
Supplier: Tonbo Biosciences
Specifications
- Antigen name:Protein Tyrosine Phosphatase Receptor Type C
- Antigen symbol:PTPRC
- Clone:HI30
- Conjugation:PerCP (Peridinin-Chlorophyll Protein Complex)-Cy5.5®
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Chimpanzee,Human
- Gene ID:5788
- Antigen synonyms:CD45R|B220|CD45|GP180|T200|L-CA|LY5|receptor type C|LCA|protein tyrosine phosphatase
- Storage temperature:Store Undiluted at 2...8 °C; Protect from Light; Do Not Freeze.
- Tested applications:Flow Cytometry
- Size:25 tests
- Pk:25 Tests
Specifications
About this item
The HI30 antibody reacts with human CD45, one of the most abundant hematopoietic markers and one that is expressed on all leukocytes (the Leukocyte Common Antigen, LCA). CD45 is a protein tyrosine phosphatase existing in several isoforms, each being generated and expressed in cell-specific patterns. With its broad cell distribution, CD45 is critical for many leukocyte functions, regulating signal transduction and cell activation associated with the T cell receptor, B cell receptor, and IL-2 receptor. Other forms of CD45, with restricted cellular expression, include CD45R (B220), CD45RA, CD45RB, CD45RO and others.
The HI30 antibody is widely used as a marker for human CD45 expression on T cells, B cells, monocytes, macrophages, and NK cells.