Anti-CD38 Mouse Antibody (violetFluor® 450) [clone: HIT2]
Supplier: Tonbo Biosciences
Certificates
About this item
The HIT2 antibody is specific for human CD38, a 45 kD type II transmembrane glycoprotein expressed on thymocytes, activated T and B cells, and monocytes as well as other non-hematopoietic cells. CD38 is an ectoenzyme that functions to catalyse the synthesis and hydrolysis of cyclic ADP-ribose and is involved in cell signaling, regulating cell adhesion, and activation. Additionally, CD38 has been shown to be a prognostic marker for some leukemias and other diseases
Type:
Antigen: CD38
Clonality:
Clone: HIT2
Conjugation: violetFluor® 450
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Specifications
- Catalog No:
- 75-0389-T100
- 75-0389-T025
- Antigen symbol:
- CD38
- CD38
- Antigen name:
- CD38 molecule
- CD38 molecule
- Antigen synonyms:
- NAD+ nucleosidase|ADP-ribosyl cyclase 1|T10|cADPr hydrolase 1|ADP-ribosyl cyclase/cyclic ADP-ribose hydrolase|cyclic ADP-ribose hydrolase 1|CD38 antigen p45
- NAD+ nucleosidase|ADP-ribosyl cyclase 1|T10|cADPr hydrolase 1|ADP-ribosyl cyclase/cyclic ADP-ribose hydrolase|cyclic ADP-ribose hydrolase 1|CD38 antigen p45
- Conjugation:
- VioletFluor® 450
- VioletFluor® 450
- Clone:
- HIT2
- HIT2
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Gene ID:
- 952
- 952
- Isotype:
- IgG1
- IgG1
- Flow cytometry:
- Yes
- Yes
- Storage temperature:
- Store undiluted at 2...8 °C, protect from light, do not freeze.
- Store undiluted at 2...8 °C, protect from light, do not freeze.
- Tested applications:
- Flow Cytometry
- Flow Cytometry