Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Streptomycin sulphate
Streptomycin sulphate, powder
Pk:
100 g
Seq:
3401
Environmentally Preferable:
:
:
1457.4 g/mol
:
3810-74-0
:
C₂₁H₃₉N₇O₁₂·1.5H₂SO₄
:
MFCD00037023