Order Entry
Export
ContactUsLinkComponent
Caffeine Standards for Liquid Chromatography
Caffeine Standards for Liquid Chromatography
Supplier:  Agilent
Certificates
undefined
Caffeine Standards for Liquid Chromatography
Supplier:  Agilent
Certificates

Specifications

  • CAS No.:
    58-08-2
  • Formula:
    C₈H₁₀N₄O₂

Specifications

Related Information