Order Entry
Export
Orders LinkContactUsLinkComponent
Potassium cyanide ≥96.0%, GPR RECTAPUR®

Potassium cyanide ≥96.0%, GPR RECTAPUR®

Corrosive
Toxic
Health
Environment
Supplier:   VWR Chemicals
Potassium cyanide ≥96.0%, GPR RECTAPUR®
Supplier:  VWR Chemicals
Certificates
Potassium cyanide ≥96.0%, GPR RECTAPUR®

3 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
  • CAS Number:151-50-8
  • Formula:KCN
  • MDL Number:MFCD00011397
  • Molecular Weight:65.12 g/mol
  • Storage Temperature:Ambient
  • EINECS:205-792-3
  • UN:1680
  • ADR:6.1,I
Specifications for Product Family

Specification Test Results

  • Appearance:
    White crystalline powder or crystals
  • Assay:
    min. 96.0%
  • Sulfide (S):
    max. 0.003%
  • Sodium (Na):
    max. 0.5%
Product Family Options

Product Information

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.