Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Lights
Accessories for Cary source lamps
Accessories for Cary source lamps
Accessories for Cary source lamps
HEWLG9820-60000
:
Agilent
Accessories for Cary source lamps
HEWLG9820-60000
:
Agilent
Description:
Loom, deuterium lamp extension, required when installing deuterium lamp mm a Cary ¹/₃/100/300 series UV-Vis
Pk:
1