Lloyd and McCown's woody plant basal salt mixture
Supplier: Apollo Scientific
Certificates
About this item
Contains macro/micronutrients as described by Lloyd and McCown, 1981. Plant tissue culture tested. Full specification given in appendix.
Contains macro/micronutrients as described by Lloyd and McCown, 1981. Plant tissue culture tested. Full specification given in appendix.