Order Entry
Export
ContactUsLinkComponent
Calcium ascorbate dihydrate ≥99%, Sigma-Aldrich®
Calcium ascorbate dihydrate ≥99%, Sigma-Aldrich®
Catalog # SIAL359645-100G
Supplier:  Merck
CAS Number:  
Calcium ascorbate dihydrate ≥99%, Sigma-Aldrich®
Catalog # SIAL359645-100G
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :426.34 g/mol
 :M-Clarity™ quality level = MQ100
CAS Number:5743-28-2
 :5743-28-21
Formula:C₁₂H₁₄CaO₁₂·2H₂O
MDL Number:MFCD00149216