Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Enzymes
Human recombinant Cytochrome P450 2D6
Human recombinant Cytochrome P450 2D6
Human recombinant Cytochrome P450 2D6
Supplier:
Apollo Scientific
Certificates
Expand image
Human recombinant Cytochrome P450 2D6
Supplier:
Apollo Scientific
About this item
Specifications
Enzyme type:
Recombinant
Species:
Human
Storage conditions:
Ambient
Enzyme name:
Cytochrome P450 2D6
Enzyme synonyms:
LKM-1
Specifications