Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Enzymes
Human recombinant AminoAcylase-1
Human recombinant AminoAcylase-1
Human recombinant AminoAcylase-1
Supplier:
Apollo Scientific
Certificates
Expand image
Human recombinant AminoAcylase-1
Supplier:
Apollo Scientific
About this item
Specifications
Enzyme type:
Recombinant
Species:
Human
Storage conditions:
Ambient
Enzyme name:
AminoAcylase-1
Enzyme synonyms:
ACY1
Specifications