Bacteria maltose phosphorylase
Catalog # APOSBIB3090-250U
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pk:1
- Species:Bacteria
- Storage conditions:Freezer
- MDL No.:MFCD03457426
- Enzyme name:Maltose phosphorylase
- Enzyme synonyms:MP, Maltose: orthophosphate 1-beta-D-glucosyltransferase
- Pk Info:250 U
Specifications
About this item
Specific activity: IU/mg protein>5 units. IU per 1ml suspension is approximately 500 units
- Ammonium sulfate suspension