Order Entry
Export
ContactUsLinkComponent
 
Anti-IFN-g Mouse Antibody
Catalog # APOSBITP1975-1MG
Supplier:  Apollo Scientific
Anti-IFN-g Mouse Antibody
Catalog # APOSBITP1975-1MG
Supplier:  Apollo Scientific

Specifications

  • Antigen name:
    Interferon-gamma
  • Antigen symbol:
    IFN-g
  • Host:
    Mouse
  • Reactivity:
    Human
  • Pk:
    1 mg

Specifications

About this item