NANOCOLOR® Photometric Tube Test Kits for Zinc
A pre-dosed NANOCOLOR® tube test for photometric determination of zinc in water.
Measurement directly in the test tube, saving time and reducing exposure to chemical reagents
- Barcode cuvette identification
- Precisely pre-dosed reagents in 16 mm (ext.Ø) tubes
The test is particularly suitable for NANOCOLOR® photometers but can be used with other photometers as well.
Sea water analysis: Yes
Hazardous material: Yes
Remark: Measuring range on NANOCOLOR VIS II
: Rugged box with 20 test tubes.
- Model:MANA985096
- Platform:NANOCOLOR® tube tests
- Storage temperature:15…25 °C
- Shelf life (months):12
- Weight:329 g (including packaging)