Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Proteins and Peptides
Human recombinant cytokeratin 18
Human recombinant cytokeratin 18
Human recombinant cytokeratin 18
Supplier:
Apollo Scientific
Certificates
Expand image
Human recombinant cytokeratin 18
Supplier:
Apollo Scientific
About this item
Specifications
Protein/peptide type:
Recombinant
Species:
Human
Protein synonyms:
CK18
Protein/peptide name:
Cytokeratin 18
Specifications