0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
ContactUsLinkComponent
Proteins and Peptides
Mai mai recombinant GH
Mai mai recombinant GH
APOSBITP1079-1MG
:
Apollo Scientific
:
Mai mai recombinant GH
APOSBITP1079-1MG
:
Apollo Scientific
:
Pk:
1 mg
Protein/peptide type:
Recombinant
Source:
Species:
Mai mai
Environmentally Preferable:
Protein synonyms:
Growth Hormone
Protein/peptide name:
GH