Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Proteins and Peptides
Human recombinant GH Pla-20K
Human recombinant GH Pla-20K
APOSBITP1069-1MG
:
Apollo Scientific
:
Human recombinant GH Pla-20K
APOSBITP1069-1MG
:
Apollo Scientific
:
Pk:
1 mg
Protein/peptide type:
Recombinant
Species:
Human
Protein synonyms:
Placental Growth Hormone-20K
Protein/peptide name:
GH Pla-20K