0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
ContactUsLinkComponent
Proteins and Peptides
Amyloid beta (1-42)
Amyloid beta (1-42)
APOSBIBA1000-1MG
:
Apollo Scientific
:
Amyloid beta (1-42)
APOSBIBA1000-1MG
:
Apollo Scientific
:
Pk:
1 mg
Seq:
23
Protein/peptide name:
Amyloid beta (1-42)
Sequence:
1-42
Trifluoroacetate salt