Order Entry
Export
ContactUsLinkComponent
NANOCOLOR® Photometric Tube Tests, Sulfide
NANOCOLOR® Photometric Tube Tests, Sulfide
Catalog # MANA985073
Supplier:  MACHEREY-NAGEL
CAS Number:  
undefined
NANOCOLOR® Photometric Tube Tests, Sulfide
Catalog # MANA985073
Supplier:  MACHEREY-NAGEL
CAS Number:  

Specifications

  • Chemical method:
    Methylene blue
  • Pk:
    1 KIT
  • Description:
    NANOCOLOR® Sulfide 3, test no. 0-73
  • Dimensions:
    160×110×119 mm (Including packaging)
  • Model:
    MANA985073
  • Platform:
    NANOCOLOR® tube tests
  • Storage temperature:
    15...25 °C
  • Shelf life (months):
    36
  • Weight:
    344 g (Including packaging)
  • For unit:
    UV/VIS II;VIS II;500 D;PF-12Plus;Advance;MT Easy VIS;MT Easy UV
  • Tests per kit:
    20
  • Range:
    0,05 - 3,00 mg/L (S²⁻)

Specifications

About this item

Photometric test kit for the determination of sulphide in water samples.

  • Measurement directly in the test tube, saving time and reducing exposure to chemical reagents
  • Bar code cuvette identification for easy test call up in NANOCOLOR® spectrophotometers
  • Precisely pre-dosed reagents with constant high quality from batch to batch
  • Colored pictogram instructions and easy handling of the tubes
  • 16 mm (OD) tubes for easy pipetting

The test kit is evaluated in 16 mm (AD) tubes. It is pre-programmed on NANOCOLOR® photometers and is also compatible with METTLER TOLEDO Easy VIS and Easy UV. Programming data for other spectrophotometers are available.

Sea water analysis: Yes

Hazardous material: Yes

Remark: Measuring range on NANOCOLOR VIS II

Sludge reagent: No

Digestion: No

NanOx metal: No

NanOx: No

Delivery information:

Rugged box with 20 test tubes. Sufficient for 20 tests.