Order Entry
Export
ContactUsLinkComponent
6-SIMA phosphoramidite
6-SIMA phosphoramidite
Catalog # APOSBIMN2046-1G
Supplier:  Apollo Scientific
CAS Number:  
6-SIMA phosphoramidite
Catalog # APOSBIMN2046-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
Storage Temperature:Ambient