3’- Amino-3’-deoxyadenosine
Catalog # APOSBIMN2044-100MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:100 mg
- Seq:2440
Specifications
About this item
:
:266.26 g/mol
Storage Temperature:Ambient
CAS Number:2504-55-4
Formula:C10H14N6O3