Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,2'-Diamino-2'-deoxyadenosine
2,2'-Diamino-2'-deoxyadenosine ≥95%
2,2'-Diamino-2'-deoxyadenosine ≥95%
2,2'-Diamino-2'-deoxyadenosine ≥95%
Supplier:
Apollo Scientific
Certificates
Expand image
2,2'-Diamino-2'-deoxyadenosine ≥95%
Supplier:
Apollo Scientific
About this item
CAS Number:
215943-79-6
:
Formula:
C₁₀H₁₅N₇O₃
MDL Number:
MFCD09750839
:
281.276 g/mol
Storage Temperature:
Ambient