9-(2-Deoxy-2-fluoro-β-D-arabinofuranosyl)adenine (FANA-A) ≥98%
Supplier: Apollo Scientific
9-(2-Deoxy-2-fluoro-β-D-arabinofuranosyl)adenine (FANA-A) ≥98%
Supplier: Apollo Scientific
Certificates
Jump to:
About this item
Synonyms: 9-(2'-Fluoro-2'-deoxy-β-D-arabinofuranosyl)adenine, 2'-FANA-A, 2'-Deoxy-2'-fluoroarabinoadenosine, (2R,3R,4S,5R)-5-(6-aminopurin-9-yl)-4-fluoro-2-(hydroxymethyl)oxolan-3-ol, (2R,3R,4S,5R)-5-(6-Amino-9H-purin-9-yl)-4-fluoro-2-(hydroxymethyl)tetrahydrofuran-3-ol, FANA-A
- CAS Number:20227-41-2
- Formula:C₁₀H₁₂FN₅O₃
- MDL Number:MFCD12911815
- Molecular Weight:269.23 g/mol
- Storage Temperature:Ambient
2 Options Available Below
Product Details & Documents
Synonyms: 9-(2'-Fluoro-2'-deoxy-β-D-arabinofuranosyl)adenine, 2'-FANA-A, 2'-Deoxy-2'-fluoroarabinoadenosine, (2R,3R,4S,5R)-5-(6-aminopurin-9-yl)-4-fluoro-2-(hydroxymethyl)oxolan-3-ol, (2R,3R,4S,5R)-5-(6-Amino-9H-purin-9-yl)-4-fluoro-2-(hydroxymethyl)tetrahydrofuran-3-ol, FANA-A
- CAS Number:20227-41-2
- Formula:C₁₀H₁₂FN₅O₃
- MDL Number:MFCD12911815
- Molecular Weight:269.23 g/mol
- Storage Temperature:Ambient



