Order Entry
Export
ContactUsLinkComponent
Carbenicillin disodium salt solution 100 mg/ml
Irritant
Health
Carbenicillin disodium salt solution 100 mg/ml
Catalog # APOSBIG1026-100ML
Supplier:  Apollo Scientific
CAS Number:  
Irritant
Health
Carbenicillin disodium salt solution 100 mg/ml
Catalog # APOSBIG1026-100ML
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 mL
  • Seq:
    2440

Specifications

About this item

 :
 :422.363 g/mol
 :GHS07
 :GHS08
Storage Temperature:Refrigerator
CAS Number:4800-94-6
Formula:C₁₇H₁₆N₂O₆S·2Na
MDL Number:MFCD00077683