NANOCOLOR® Photometric Tube Test Kits for Hardness
A range of pre-dosed NANOCOLOR® tube tests for photometric determination of water hardness.
Measurement directly in the test tube, saving time and reducing exposure to chemical reagents
- Barcode cuvette identification
- Precisely pre-dosed reagents in 16 mm (ext.Ø) tubes
The test kits are evaluated in 16 mm (ext.Ø) tubes. They are preprogrammend on NANOCOLOR® photometers and also compatible with METTLER TOLEDO Easy VIS and Easy UV. Programming data for other spectrophotometers are available.
Sea water analysis: Yes
Hazardous material: Yes
Remark: Measuring range on NANOCOLOR VIS II
Sludge reagent set: No
Digestion set: No
NanOx Metal: No
NanOx N: No
: Remark: Measuring range on NANOCOLOR VIS II
- Model:MANA985044
- Platform:NANOCOLOR® tube tests
- Storage temperature:15…25 °C
- Shelf life (months):18
- Weight:463 g (including packaging)