Patulin standard solution
Supplier: Apollo Scientific
About this item
Analytical reference standard solution in acetonitrile.
Caution: Expires in 12 months.
2 Options Available Below
Product Details & Documents
Analytical reference standard solution in acetonitrile.
Caution: Expires in 12 months.
Documents
Certificates
Search by Lot NumberSpecifications for Product Family
Specifications
Specifications
- Catalog No:
- APOSBIMS108-1ML
- APOSBIMS108-5ML
- Description:
- Patulin
- Patulin
- CAS No.:
- 149-29-1
- Formula:
- C₇H₆O₄
- Boiling point:
- 81…82 °C
- Flash point:
- 2 °C
- Storage temperature:
- Freezer
Specifications
Product Family Options
Product Information
- ConcentrationPack typePkAvailabilityPrice
- APOSBIMS108-1MLPatulinSpecifications:
More
Specifications
Cat. No.APOSBIMS108-1MLDescriptionPatulinCAS No.149-29-1FormulaC₇H₆O₄Boiling point81…82 °CFlash point2 °CStorage temperatureFreezer - APOSBIMS108-5MLPatulinSpecifications:
More
Specifications
Cat. No.APOSBIMS108-5MLDescriptionPatulinCAS No.FormulaBoiling pointFlash pointStorage temperature



