
2,6-Difluoro-4-hydroxybenzaldehyde 98%
Catalog # APOSPC9940-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:158.1 g/mol
Melting Pt:179…182 °C
:GHS07
Storage Temperature:Ambient
CAS Number:532967-21-8
Formula:C₇H₄F₂O₂
MDL Number:MFCD07772042